• USA Home
  • 371408 - 5,5-Dimethyl-1,3-dioxane-2-ethanol

EMAIL THIS PAGE TO A FRIEND
371408 Aldrich

5,5-Dimethyl-1,3-dioxane-2-ethanol

99%

Purchase

Properties

Related Categories Alcohols, Building Blocks, C7 to C8, Chemical Synthesis, Organic Building Blocks,
InChI Key   HYGOOWUTSBFURW-UHFFFAOYSA-N
assay   99%
refractive index   n20/D 1.45(lit.)
density   1.031 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
224.6 °F
Flash Point(C) 
107 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?