EMAIL THIS PAGE TO A FRIEND
372390 Aldrich

Lithium

99%, high sodium

Purchase

Properties

InChI Key   WHXSMMKQMYFTQS-UHFFFAOYSA-N
assay   99%
form   powder
resistivity   9.446 μΩ-cm, 20°C
impurities   0.5% sodium
bp   1342 °C(lit.)
mp   180 °C(lit.)
density   0.534 g/mL at 25 °C(lit.)

Description

Packaging

Packaged under argon

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 1415 4.3 / PGI
WGK Germany 
2
RTECS 
OJ5540000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?