EMAIL THIS PAGE TO A FRIEND
373222 Aldrich

Platinum

evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis

Purchase

Properties

Related Categories Evaporation Slugs, Materials Science, Metal and Ceramic Science, Metals, Micro/NanoElectronics,
InChI Key   BASFCYQUMIYNBI-UHFFFAOYSA-N
assay   99.99% trace metals basis
form   evaporation slug
resistivity   10.6 μΩ-cm, 20°C
diam. × L   0.6 cm × 0.6 cm
weight   3.6 g
bp   3827 °C(lit.)
mp   1772 °C(lit.)
density   21.45 g/cm3(lit.)

Price and Availability


Ultra-High Purity Inorganics
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles

Articles

Combinatorial Materials Science for Energy Applications

Materials are the fundamental basis for solutions to the most pressing issues in energy generation, transport, and utilization, as well as more general issues in sustainability. In many cases, long-t...
Keywords: Asymmetric synthesis, Catalysis, Chemical vapor deposition, Combinatorial synthesis, Dehydrogenation, Deposition, Diffraction, Diffusion, Evaporation, Genetic, Hydration reaction, Materials Science, Microscopy, Nucleic acid annealing, Oxidations, Photovoltaics, Polymerization reactions, Reductions, Semiconductor, Substitutions, Thin film deposition, X-Ray diffraction

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?