CAS Number 69991-67-9
Linear Formula [CF(CF3)CF2O]x(CF2O)y
MDL number MFCD00134390
PubChem Substance ID 24863310
Popular Documents: FTIR (PDF)
| feed ratio | 30:1 (x:y) |
| mol wt | average Mn ~6,600 |
| refractive index |
n |
| density | 1.892 g/mL at 25 °C |
High performance lubricant for textile, bearing, and corrosive environment applications.
Polymer is made by an oxidative polymerization of hexafluoropropylene and tetrafluoroethylene.
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?