• USA Home
  • 374865 - 2-Chloro-5-(trifluoromethyl)phenyl isocyanate

EMAIL THIS PAGE TO A FRIEND
374865 Aldrich

2-Chloro-5-(trifluoromethyl)phenyl isocyanate

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Isocyanates, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   WEPYOPYMWSHRIW-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.489(lit.)
bp   63-64 °C/5 mmHg(lit.)
density   1.48 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2206PSN1B 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
197.6 °F
Flash Point(C) 
92 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?