EMAIL THIS PAGE TO A FRIEND
375675 Aldrich

Triisodecylamine

technical grade, 90%

Purchase

Properties

Related Categories Amines, Building Blocks, C20 to C30+, Chemical Synthesis, Nitrogen Compounds,
grade   technical grade
InChI Key   DLFKJPZBBCZWOO-UHFFFAOYSA-N
assay   90%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?