• USA Home
  • 376809 - S-Phenyl thiopropionate

EMAIL THIS PAGE TO A FRIEND

376809 Aldrich

S-Phenyl thiopropionate

98%

Purchase

Properties

assay   98%
refractive index   n20/D 1.56(lit.)
bp   130-131 °C/20 mmHg(lit.)
density   1.083 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
Xn
Risk Statements (Europe) 
Safety Statements (Europe) 
26-37/39
RIDADR 
UN 3334
WGK Germany 
3
Flash Point(F) 
228.2 °F
Flash Point(C) 
109 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

FT-NMR 1 (2), 1286:A / RegBook 1 (2), 1927:C / Structure Index 1, 309:B:7

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?