• USA Home
  • 37690 - Dihydroxyfumaric acid dihydrate

EMAIL THIS PAGE TO A FRIEND
37690 Aldrich

Dihydroxyfumaric acid dihydrate

technical, ≥90% (T)

  • CAS Number 20688-70-4

  • Empirical Formula (Hill Notation) C4H4O6 · 2H2O

  • Molecular Weight 184.10

  •  Beilstein Registry Number 1724790

  •  MDL number MFCD00149287

  •  PubChem Substance ID 57649745

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   LDRMCGGUXOJSDA-SEPHDYHBSA-N
grade   technical
assay   ≥90% (T)
mp   ~155 °C (dec.)
solubility   methanol: soluble 1 g/10 mL clear, colorless (hot)
storage temp.   2-8°C

Description

Packaging

10 g in glass bottle

Application

Dihydroxyfumaric acid dihydrate may be used in the preparation of mesoxalic acid semialdehyde and its derivatives.

General description

Crystal structure of dihydroxyfumaric acid dihydrate has been reported. Homogeneous reaction of dihydroxyfumarate (DHF) with glyoxylic acid monohydrate has been reported. It is also reported to react with glyoxylate, glycolaldehyde and glyceraldehyde, via α-keto acid intermediates, to afford triose, tetrulose and pentuloses.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?