EMAIL THIS PAGE TO A FRIEND
378003 Aldrich

3-Ethylpyridine

technical grade, 94%

  • CAS Number 536-78-7

  • Empirical Formula (Hill Notation) C7H9N

  • Molecular Weight 107.15

  •  Beilstein Registry Number 106479

  •  EC Number 208-647-2

  •  MDL number MFCD00006413

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

grade   technical grade
InChI Key   MFEIKQPHQINPRI-UHFFFAOYSA-N
assay   94%
impurities   6% 2,3-dimethylpyridine
refractive index   n20/D 1.502(lit.)
bp   163-166 °C(lit.)
density   0.954 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2924 8(3) / PGIII
WGK Germany 
3
Flash Point(F) 
123.8 °F
Flash Point(C) 
51 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?