• USA Home
  • 381705 - 2-Amino-3,4,5,6-tetrafluorobenzoic acid

EMAIL THIS PAGE TO A FRIEND
381705 Aldrich

2-Amino-3,4,5,6-tetrafluorobenzoic acid

98%

Synonym: 3,4,5,6-Tetrafluoroanthranilic acid

Purchase

Properties

Related Categories Aromatic Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   CNSGPAMLXMBLNA-UHFFFAOYSA-N
assay   98%
mp   141-143 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?