• USA Home
  • 382469 - O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N′,N′-tetramethyluronium tetrafluoroborate

EMAIL THIS PAGE TO A FRIEND
382469 Aldrich

O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N′,N′-tetramethyluronium tetrafluoroborate

98%

Synonym: N-[[[(1-Cyano-2-ethoxy-2-oxoethylidene)amino]oxy](dimethylamino)methylene]-N-methyl-methanaminium tetrafluoroborate, TOTU

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Coupling, Peptide Chemistry, Peptide Coupling,
InChI Key   FPQVGDGSRVMNMR-JCTPKUEWSA-N
assay   98%
mp   145 °C (dec.)(lit.)
storage temp.   2-8°C

Description

Application

Reactant for:
Watson-Crick pairing of oligonucleotides
Esterification of nucleosides to solid phase supports
Reagent for:
Synthesis of selective integrin avβ3 antagonists
Design and synthesis of double stranded DNA libraries
Preparation of conformationally contrained β-amino acid derivatives
Synthesis and application of methyleneoxy pseudopeptide building blocks

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

COMU – Safer and More Efficient Peptide Coupling Reagent

Peptide synthesis relies heavily on efficient and reliable coupling reagents. A low tendency for racemization is a key requirement. This is especially true for solid phase peptide synthesis—quantitat...
Keywords: Building blocks, Calorimetry, Catalog, Condensations, Coupling reactions, Epimerizations, Methods, PEGylations, Peptide synthesis, Protocols, Racemizations, Search, Solid phase peptide synthesis, Solvents, Tools

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?