• USA Home
  • 382531 - N,N-Dimethylundecylamine N-oxide

EMAIL THIS PAGE TO A FRIEND
382531 Aldrich

N,N-Dimethylundecylamine N-oxide

97%

  • CAS Number 15178-71-9

  • Linear Formula CH3(CH2)10N(O)(CH3)2

  • Molecular Weight 215.38

  •  Beilstein Registry Number 2355011

  •  MDL number MFCD00144984

  •  PubChem Substance ID 24863903

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Amine Oxides, Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   OZHBUVQCJMARBN-UHFFFAOYSA-N
assay   97%
mp   129-131 °C(lit.)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?