• USA Home
  • 383635 - 5-Methyl-3-(methylthio)-1,4-diphenyl-1H-1,2,4-triazolium iodide

EMAIL THIS PAGE TO A FRIEND
383635 Aldrich

5-Methyl-3-(methylthio)-1,4-diphenyl-1H-1,2,4-triazolium iodide

98%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Triazoles
InChI Key   DYIKGUMVIOCUSG-UHFFFAOYSA-M
assay   98%
mp   254-255 °C(lit.)

Description

Packaging

5 g in glass bottle

General description

5-Methyl-3-(methylthio)-1,4-diphenyl-1H-1,2,4-triazolium iodide is a heterocyclic building block. It is a triazole derivative.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?