• USA Home
  • 384135 - Trichloro[1,1,1-tris(diphenylphosphinomethyl)ethane]rhodium(III)

EMAIL THIS PAGE TO A FRIEND
384135 Aldrich

Trichloro[1,1,1-tris(diphenylphosphinomethyl)ethane]rhodium(III)

Synonym: [1,1,1-Tris(diphenylphosphinomethyl)ethane]rhodium(III) chloride

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Rhodium More...
InChI Key   ZAUZIYFSWQXYJZ-UHFFFAOYSA-K
mp   312 °C (dec.)(lit.)

Description

Packaging

500 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?