• USA Home
  • 386170 - 2-{2-[(3-Ethyl-5-methyl-2(3H)-benzothiazolylidene)methyl]-1-butenyl}-1-(3-sulfopropyl)naphtho[1,2-d]thiazolium hydroxide inner salt

EMAIL THIS PAGE TO A FRIEND
386170 Aldrich

2-{2-[(3-Ethyl-5-methyl-2(3H)-benzothiazolylidene)methyl]-1-butenyl}-1-(3-sulfopropyl)naphtho[1,2-d]thiazolium hydroxide inner salt

Dye content 85 %

Purchase

Properties

Related Categories Cell Biology, E-F, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   BCJMYXULAHGVFY-UHFFFAOYSA-N
composition   Dye content, 85%
mp   192 °C (dec.)(lit.)
absorption   λmax 566 nm

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?