• USA Home
  • 387525 - Dihydro-5-[5-(1,2,3,4-tetrahydro-6-hydroxy-4-oxo-2-thioxo-5-pyrimidinyl)-2,4-pentadienylidene]-2-thioxo-4,6(1H,5H)pyrimidinedione

EMAIL THIS PAGE TO A FRIEND
387525 Aldrich

Dihydro-5-[5-(1,2,3,4-tetrahydro-6-hydroxy-4-oxo-2-thioxo-5-pyrimidinyl)-2,4-pentadienylidene]-2-thioxo-4,6(1H,5H)pyrimidinedione

Dye content 90 %

  • CAS Number 63811-40-5

  • Empirical Formula (Hill Notation) C13H10N4O4S2

  • Molecular Weight 350.37

  •  PubChem Substance ID 329755780

Purchase

Properties

InChI Key   LFYHUCUXVULCOB-ZPUQHVIOSA-N
composition   Dye content, 90%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?