• USA Home
  • 388599 - 4,5-Dimethoxy-2-nitrobenzonitrile

EMAIL THIS PAGE TO A FRIEND
388599 Aldrich

4,5-Dimethoxy-2-nitrobenzonitrile

99%

Purchase

Properties

InChI Key   NQSQQGDTYKGCOT-UHFFFAOYSA-N
assay   99%
mp   164-168 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

650102 4,5-Dimethoxy-2-nitrobenzonitrile, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?