EMAIL THIS PAGE TO A FRIEND
388653 Aldrich

Polybutenes

average Mn ~560 by VPO, isobutylene >90 %

Purchase

Properties

InChI Key   WTOOLIQYCQJDBG-BJILWQEISA-N
mol wt   average Mn ~560 by VPO
composition   isobutylene, >90%
viscosity   29-35 cSt(99 °C)(lit.)
transition temp   pour point (ASTM D 97) −35 °C

Description

Application

Hydrophobic material is derivatized to make detergents and anti-rust additives. Adds tack, flexibility and water repellency to adhesives and coatings. Contributes ′cling′ to LLDPE films. Plasticizer for polymers, lubricants and metal-working fluids.

Features and Benefits

Tackiness increases with molecular weight. Some internal double bonds may be present.

Other Notes

remainder 1- and 2-butenes

Physical form

Terminal vinyl group.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
1
RTECS 
EM9032000
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?