• USA Home
  • 388793 - 3-(Decyldimethylammonio)propanesulfonate inner salt

EMAIL THIS PAGE TO A FRIEND
388793 Aldrich

3-(Decyldimethylammonio)propanesulfonate inner salt

97%

Synonym: Caprylyl sulfobetaine, SB3-10, Zwittergent® 3-10

  • CAS Number 15163-36-7

  • Linear Formula CH3(CH2)9N+(CH3)2CH2CH2CH2SO3-

  • Molecular Weight 307.49

  •  Beilstein Registry Number 3967284

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

InChI Key   MKMQVUIGDCFLFL-UHFFFAOYSA-N
assay   97%

Description

Legal Information

Zwittergent is a registered trademark of EMD BioScience, Inc.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?