EMAIL THIS PAGE TO A FRIEND
388955 Aldrich

Cetrimide

Synonym: Alkyltrimethylammonium bromide, predominantly C14

Purchase

Properties

mp   239 °C (dec.)(lit.)

Description

Other Notes

This product has been replaced by M7635-SIGMA | Alkyltrimethylammonium bromide ≥95% (TLC)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3263 8 / PGII
WGK Germany 
3
RTECS 
FL2797500
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

M7635 Alkyltrimethylammonium bromide, ≥95% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?