• USA Home
  • 390291 - Copper(I) 1-butanethiolate

EMAIL THIS PAGE TO A FRIEND
390291 Aldrich

Copper(I) 1-butanethiolate

Synonym: 1-Butanethiol copper(I) salt

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiol/Mercaptan Salts More...
InChI Key   WJXDCSSGTLPZDK-UHFFFAOYSA-M

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3335
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?