• USA Home
  • 390992 - 2,4,6-Tri-tert-butyl-N-methylaniline

EMAIL THIS PAGE TO A FRIEND
390992 Aldrich

2,4,6-Tri-tert-butyl-N-methylaniline

95%

Purchase

Properties

Related Categories Amines, Building Blocks, C15 to C19, Chemical Synthesis, Nitrogen Compounds,
InChI Key   GFTNLYGZUUPSSM-UHFFFAOYSA-N
assay   95%
mp   109-111 °C(lit.)

Description

Packaging

1 g in glass bottle

250 mg in glass bottle

Application

2,4,6-Tri-tert-butyl-N-methylaniline may be used in the synthesis of 2,5-di-tert-butyl-N,N-dimethylanilinium iodide and 2,5-di-tert-butyl-N-ethyl-N-ethylanilinium iodide.

General description

2,4,6-Tri-tert-butyl-N-methylaniline, a sterically hinderd amine, is an aniline derivative. The steric effect on charge-transfer (CT) band in its electronic spectra has been reported. The reactivity of 2,4,6-tri-tert-butyl-N-methylaniline with alkyl iodides under high pressure has been reported.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?