• USA Home
  • 392782 - 3,5-Diacetyl-2,6-dimethylpyridine

EMAIL THIS PAGE TO A FRIEND
392782 Aldrich

3,5-Diacetyl-2,6-dimethylpyridine

98%

Purchase

Properties

Related Categories Building Blocks, C11, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   RCACBGGDSKMOCT-UHFFFAOYSA-N
assay   98%
mp   71-73 °C(lit.)

Description

Application

3,5-Diacetyl-2,6-dimethylpyridine may be used the synthesis of the following:
• substituted azachalcone derivatives
• bis-β-diketones and difluoroboron bis-β-diketonate derivatives
• (1-acetyl-5-aryl-4,5-dihydro)-1H-pyrazole substituted pyridine derivatives

General description

3,5-Diacetyl-2,6-dimethylpyridine is a diacetyl pyridine derivative. It has been reported to be formed by the photooxidation of 3,5-diacetyl-1,4-dihydro-2,6-dimethylpyridine.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?