• USA Home
  • 39398 - Alloc-Phe-OH (dicyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
39398 Aldrich

Alloc-Phe-OH (dicyclohexylammonium) salt

≥97.0%

Synonym: Alloc-L-phenylalanine (dicyclohexylammonium) salt, N-Allyloxycarbonyl-L-phenylalanine (dicyclohexylammonium) salt

  • CAS Number 110637-43-9

  • Empirical Formula (Hill Notation) C13H15NO4

  • Molecular Weight 249.26

  •  Beilstein Registry Number 4633229

  •  MDL number MFCD04973097

  •  PubChem Substance ID 57649896

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylalanine More...
InChI Key   BUWUDPXYWHZZKS-MERQFXBCSA-N
assay   ≥97.0%

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?