• USA Home
  • 394092 - 1-(4-Nitrophenyl)-3-pyrrolidino-2-pyrazolin-5-one

EMAIL THIS PAGE TO A FRIEND
394092 Aldrich

1-(4-Nitrophenyl)-3-pyrrolidino-2-pyrazolin-5-one

95%

Purchase

Properties

Related Categories Building Blocks, C13 to C20+, Chemical Synthesis, Heterocyclic Building Blocks, Pyrrolidines More...
InChI Key   GMXRVALYFNKUTQ-UHFFFAOYSA-N
assay   95%
mp   205-208 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?