• USA Home
  • 394939 - N-Methyl-bis(trifluoroacetamide)

EMAIL THIS PAGE TO A FRIEND
394939 Aldrich

N-Methyl-bis(trifluoroacetamide)

derivatization grade, 97%

Synonym: MBTFA

Purchase

Properties

grade   derivatization grade
InChI Key   AWGBWLXGUPTXHF-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.346(lit.)
bp   121-122 °C(lit.)
density   1.547 g/mL at 25 °C

Description

Application

Reagent for introducing trifluoroacetyl group.

Trifluoroacetylating reagent for alcohols, thiols, phenols, carboxylic acids, amines, and amides.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2920 3(8) / PGII
WGK Germany 
3
Flash Point(F) 
113 °F
Flash Point(C) 
45 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M0789 N-Methyl-bis(trifluoroacetamide), for GC derivatization, ≥97.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?