EMAIL THIS PAGE TO A FRIEND
395323 Aldrich

Alkali Blue 6B

Dye content 50 %

Synonym: Acid Blue 110, Acid Blue 119

  • CAS Number 1324-76-1

  • Empirical Formula (Hill Notation) C37H31N3O4S

  • Molecular Weight 613.72

  •  Colour Index Number 42750

  •  EC Number 250-253-8

Purchase

Properties

composition   Dye content, 50%

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DB4957000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?