• USA Home
  • 396125 - Methacryloyl isocyanate

EMAIL THIS PAGE TO A FRIEND
396125 Aldrich

Methacryloyl isocyanate

technical grade, 90%

  • CAS Number 4474-60-6

  • Empirical Formula (Hill Notation) C5H5NO2

  • Molecular Weight 111.10

  •  PubChem Substance ID 329755887

Purchase

Properties

grade   technical grade
InChI Key   JEHFRMABGJJCPF-UHFFFAOYSA-N
assay   90%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?