• USA Home
  • 398772 - N,N-Dimethyloctylamine

EMAIL THIS PAGE TO A FRIEND
398772 Aldrich

N,N-Dimethyloctylamine

technical grade

Synonym: 1-(Dimethylamino)octane, N-Octyldimethylamine, Adma 8, DMOA

Purchase

Properties

Related Categories Amines, Building Blocks, C10, Chemical Synthesis, Nitrogen Compounds,
grade   technical grade
vapor density   5.4 (vs air)
vapor pressure   0.8 mmHg ( 25 °C)
InChI Key   UQKAOOAFEFCDGT-UHFFFAOYSA-N
refractive index   n20/D 1.424(lit.)
bp   195 °C(lit.)
mp   −57 °C(lit.)
density   0.765 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2922 6.1(8) / PGIII
WGK Germany 
1
RTECS 
RG8075000
Flash Point(F) 
147.2 °F
Flash Point(C) 
64 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

256226 N,N-Dimethyloctylamine, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?