CAS Number 70456-74-5
Linear Formula NH4F · HF
MDL number MFCD03096499
| Related Categories | Electronic Chemicals, Etchants, Materials Science, Micro/NanoElectronics |
| grade | semiconductor grade PURANAL™ |
| CofA | specification on request |
PURANAL is a trademark of Honeywell Specialty Chemicals Seelze GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?