40213 Aldrich

Hydrofluoric acid

semiconductor grade PURANAL (Honeywell 17736)

DOWNLOAD MSDS (PDF)

Synonym: HF

  • CAS Number 7664-39-3

  • Empirical Formula (Hill Notation) HF

  • Molecular Weight 20.01

  •  EC Number 231-634-8

  •  MDL number MFCD00011346

  •  PubChem Substance ID 24865049

Properties

Related Categories Chemical Synthesis, Electronic Chemicals, Inorganic Acids, Materials Science, Micro/NanoElectronics,
grade   semiconductor grade PURANAL (Honeywell 17736)
vapor density   1.27 (vs air)
vapor pressure   25 mmHg ( 20 °C)
CofA   specification on request
density   1.15 g/mL at 25 °C(lit.)

Description

Legal Information

PURANAL is a trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Customers Also Viewed

Ammonium fluoride – hydrofluoric acid mixture

AF 875-125, etching mixture, semiconductor grade PURANAL

Hydrofluoric acid

semiconductor grade PURANAL (Honeywell 17735)

Hydrofluoric acid

semiconductor grade MOS PURANAL (Honeywell 17928)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?