• USA Home
  • 40235 - (−)-N,N-Dimethylephedrinium bromide

EMAIL THIS PAGE TO A FRIEND
40235 Aldrich

(−)-N,N-Dimethylephedrinium bromide

≥99.0% (AT)

  • CAS Number 55380-59-1

  • Empirical Formula (Hill Notation) C12H20BrNO

  • Molecular Weight 274.20

  •  Beilstein Registry Number 6099406

  •  EC Number 259-619-1

  •  MDL number MFCD00043190

  •  PubChem Substance ID 57649947

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Phase Transfer Reaction
InChI Key   QNYXXJBMEPUNIY-JGAZGGJJSA-M
assay   ≥99.0% (AT)
optical activity   [α]20/D −39±1°, c = 5% in methanol
mp   236-238 °C

Description

Other Notes

Chiral phase-transfer catalyst. Review; Chiral supporting electrolyte; For the diastereoselective hydration of prochiral C-C double bonds

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?