• USA Home
  • 40238 - 2-Fluoro-1,3-dimethylpyridinium p-toluenesulfonate

EMAIL THIS PAGE TO A FRIEND
40238 Aldrich

2-Fluoro-1,3-dimethylpyridinium p-toluenesulfonate

≥95.0% (N)

Synonym: 1,3-Dimethyl-2-fluoropyridinium p-toluenesulfonate

Purchase

Properties

Related Categories Building Blocks, C7, Chemical Synthesis, Fluorination, Fluorination Reagents,
InChI Key   HXUOPSAHPZTNGS-UHFFFAOYSA-M
assay   ≥95.0% (N)
mp   111-116 °C
storage temp.   2-8°C

Description

Other Notes

Reagent for the efficient preparation of carboxylic acid fluorides, the cyclisation of unsaturated alcohols and a convenient method for the oxidation of alcohols. Review

Packaging

5 g in glass bottle

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?