• USA Home
  • 402591 - 1,4-Dichloroanthraquinone

EMAIL THIS PAGE TO A FRIEND
402591 Aldrich

1,4-Dichloroanthraquinone

97%

  • CAS Number 602-25-5

  • Empirical Formula (Hill Notation) C14H6Cl2O2

  • Molecular Weight 277.10

  •  MDL number MFCD00192002

  •  PubChem Substance ID 24865102

Purchase

Properties

Related Categories Building Blocks, C13 to C14, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   CAHGWVAXFJXDNI-UHFFFAOYSA-N
assay   97%
mp   187-189 °C(lit.)

Description

Application

1,4-Dichloroanthraquinone may be employed in the synthesis of synthesis of a series of 1-alkylaminoanthraquinones.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?