• USA Home
  • 40265 - Tetramethylammonium hydroxide solution

EMAIL THIS PAGE TO A FRIEND
40265 Aldrich

Tetramethylammonium hydroxide solution

semiconductor grade PURANAL (Honeywell 17741)

  • CAS Number 75-59-2

  • Linear Formula (CH3)4N(OH)

  • Molecular Weight 91.15

  •  Beilstein Registry Number 3558708

  •  MDL number MFCD00008280

  •  PubChem Substance ID 57649955

Purchase

Properties

Related Categories Electronic Chemicals, Materials Science, Micro/NanoElectronics, Semiconductor Grade Chemicals
grade   semiconductor grade PURANAL (Honeywell 17741)
InChI Key   WGTYBPLFGIVFAS-UHFFFAOYSA-M
CofA   specification on request

Description

Legal Information

PURANAL is a trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1835 8 / PGII
WGK Germany 
3
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?