EMAIL THIS PAGE TO A FRIEND
40303 Aldrich

Nitric acid

semiconductor grade VLSI PURANAL (Honeywell 17512), 69-71%

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Electronic Chemicals, Inorganic Acids,
grade   semiconductor grade VLSI PURANAL (Honeywell 17512)
vapor pressure   8 mmHg ( 20 °C)
InChI Key   GRYLNZFGIOXLOG-UHFFFAOYSA-N
assay   69-71%
CofA   specification on request
color   colorless
bp   120.5 °C(lit.)
density   1.400 g/cm3 at 25 °C

Description

Legal Information

PURANAL is a trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Corrosive to the respiratory tract.
RIDADR 
UN2031 - class 8 (5.1) - PG 2 - IATA PAX forbidden - Nitric acid >=65% - <= 70%
WGK Germany 
2
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?