• USA Home
  • 404128 - Calcium AA/ICP calibration/check standard for environmental analysis

EMAIL THIS PAGE TO A FRIEND
404128 Aldrich

Calcium AA/ICP calibration/check standard for environmental analysis

1,000 μg/mL Ca+2 in 2% HNO3 (Exact Ca concentration on label)

Purchase

Properties

Related Categories AA/ICP Standard Solutions, AAS, Additional Standards, Analytical Standards, Analytical/Chromatography,
concentration   1,000 μg/mL Ca+2 in 2% HNO3 (Exact Ca concentration on label)
application(s)   ICP/AAS: suitable
density   1.01 g/mL at 25 °C
format   single component solution

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3264 8/PG 3
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

19051 Calcium Standard for ICP, TraceCERT®, 1000 mg/L Ca in nitric acid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?