EMAIL THIS PAGE TO A FRIEND
405183 Aldrich

IR-1050

Dye content 97 %

Synonym: 1-Butyl-2-[2-[3-[(1-butyl-6-chlorobenz[cd]indol-2(1H)-ylidene)ethylidene]-2-chloro-5-methyl-1-cyclohexen-1-yl]ethenyl]-6-chlorobenz[cd]indolium tetrafluoroborate

  • CAS Number 155614-00-9

  • Empirical Formula (Hill Notation) C41H40BCl3F4N2

  • Molecular Weight 753.93

  •  MDL number MFCD00191875

  •  PubChem Substance ID 24865297

Purchase

Properties

Related Categories Infrared (IR) Dyes, Laser Dyes, Materials Science, Organic and Printed Electronics, Photonic and Optical Materials More...
InChI Key   MIQPSXMNDUZFOT-UHFFFAOYSA-N
composition   Dye content, 97%
mp   233-235 °C(lit.)
absorption   λmax 1048 nm in ethanol

Description

Features and Benefits

Q-switching IR laser dye.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?