• USA Home
  • 405469 - Poly(Bisphenol A-co-epichlorohydrin), glycidyl end-capped

EMAIL THIS PAGE TO A FRIEND
405469 Aldrich

Poly(Bisphenol A-co-epichlorohydrin), glycidyl end-capped

average Mn ~4,000

Purchase

Properties

Related Categories Bisphenol A Polymers and Epoxy Prepolymer Resins, Engineering Polymers, Materials Science, Polymer Science, Polymers More...
mol wt   average Mn ~4,000
mp   120-130 °C
transition temp   Tg (DSC) 75 °C (onset)

Price and Availability


Webinar: Recent Developments in RAFT Agents
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?