• USA Home
  • 406376 - Isoamyl 4-(dimethylamino)benzoate,mixture of isomers

EMAIL THIS PAGE TO A FRIEND
406376 Aldrich

Isoamyl 4-(dimethylamino)benzoate,mixture of isomers

98%

Purchase

Properties

Related Categories Amine Photochemical Coinitiators, Materials Science, Polymer Science, Polymerization Initiators, Reagents for Polymerization More...
InChI Key   OFSAUHSCHWRZKM-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.553(lit.)
bp   150-155 °C/2.5 mmHg(lit.)
density   1.02 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
185 °F
Flash Point(C) 
85 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?