• USA Home
  • 406589 - Poly[(phenyl isocyanate)-co-formaldehyde]

EMAIL THIS PAGE TO A FRIEND
406589 Aldrich

Poly[(phenyl isocyanate)-co-formaldehyde]

average Mn ~290

Purchase

Properties

activity   ~2.3 isocyanate groups/molecule
mol wt   average Mn ~290
viscosity   50 cP(25 °C)(lit.)

Description

Application

Films and foams for coatings, adhesives, packaging, furniture and auto interiors.

Features and Benefits

Low viscosity, low functionality, high dimer content and high percentage of ortho and para isomers.

Physical form

Oligomer.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3334
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?