• USA Home
  • 406635 - Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol) bis(2-aminopropyl ether)

EMAIL THIS PAGE TO A FRIEND
406635 Aldrich

Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol) bis(2-aminopropyl ether)

average Mn ~2,000

Synonym: Jeffamine® XTJ-502

  • CAS Number 65605-36-9

  • Linear Formula CH3CH(NH2)CH2[OCH(CH3)CH2]l(OCH2CH2)m[OCH2CH(CH3)]nNH2

  •  MDL number MFCD00197920

  •  PubChem Substance ID 24865394

Purchase

Description

Legal Information

Jeffamine is a registered trademark of Huntsman Petrochemical, LLC

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?