CAS Number 6891-44-7
Linear Formula H2C=C(CH3)CO2CH2CH2N(CH3)3(OSO3CH3)
Molecular Weight 283.34
MDL number MFCD00078263
PubChem Substance ID 24865506
| Related Categories | Acrylic Monomers, Materials Science, Methacrylate, Monomers, Polymer Science More... |
| contains | 900 ppm monomethyl ether hydroquinone as inhibitor |
| concentration | 80 wt. % in H2O |
| refractive index |
n |
| density | 1.183 g/mL at 25 °C |
500 mL in glass bottle
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?