• USA Home
  • 40876 - (+)-2,3-O-Benzylidene-1,1,4,4-tetraphenyl-D-threitol polymer-bound

EMAIL THIS PAGE TO A FRIEND
40876 Aldrich

(+)-2,3-O-Benzylidene-1,1,4,4-tetraphenyl-D-threitol polymer-bound

extent of labeling: ~0.40 mmol/g loading

Synonym: (4S,5S)-2,α,α,α′,α′-Pentaphenyl-dioxolane-4,5-dimethanol polymer-bound, (S,S)-Taddol (phenyl) polymer-bound

Purchase

Properties

Related Categories Chemical Synthesis, Other Supported Reagents, Solid Supported Synthesis, Specialty Synthesis, Supported Reagents More...
extent of labeling   ~0.40 mmol/g loading

Description

Other Notes

Chiral supported ligand; used in the preparation of a chiral titanium catalyst employed for asymmetric additions of alkyl groups to aldehydes

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?