• USA Home
  • 410098 - Tetra(ethylene glycol) divinyl ether

EMAIL THIS PAGE TO A FRIEND
410098 Aldrich

Tetra(ethylene glycol) divinyl ether

98%

  • CAS Number 83416-06-2

  • Linear Formula (H2C=CHOCH2CH2OCH2CH2)2O

  • Molecular Weight 246.30

  •  MDL number MFCD00192471

Purchase

Properties

InChI Key   UEIPWOFSKAZYJO-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.456(lit.)
bp   102 °C/0.15 mmHg(lit.)
density   1.03 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

329800 Tri(ethylene glycol) divinyl ether, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?