EMAIL THIS PAGE TO A FRIEND
411671 Aldrich

Paraffin wax

mp 70-80 °C (ASTM D 127)

Purchase

Properties

Related Categories Engineering Polymers, Materials Science, Polymer Science, Polymers, Waxes and Oils More...
mp   70-80 °C (ASTM D 127)

Price and Availability


Laboratory Gloves

Webinar: Recent Developments in RAFT Agents
Safety & Documentation

Safety Information

WGK Germany 
nwg
RTECS 
RV0350000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?