• USA Home
  • 415030 - N-(2-Furfurylideneacetyl)glycine methyl ester

EMAIL THIS PAGE TO A FRIEND
415030 Aldrich

N-(2-Furfurylideneacetyl)glycine methyl ester

98%

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Glycine Derivatives, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   BHKITARSEBNDBI-SNAWJCMRSA-N
assay   98%
mp   133-136 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?