• USA Home
  • 416304 - Poly(methyl vinyl ether-alt-maleic acid monobutyl ester) solution

EMAIL THIS PAGE TO A FRIEND
416304 Aldrich

Poly(methyl vinyl ether-alt-maleic acid monobutyl ester) solution

average Mw ~135,000 by LS, 50 wt. % in ethanol

  • CAS Number 25119-68-0

  • Linear Formula [CH2CH(OCH3)CH(CO2R)CH(CO2R)]n, R = H or -CH2CH2CH2CH3

  •  MDL number MFCD00197915

  •  PubChem Substance ID 24866068

Purchase

Properties

Related Categories Hydrophilic Polymers, Maleic Anhydride Copolymers, Materials Science, Miscellaneous Hydrophilic Polymers, Poly(methyl vinyl ether) and Copolymers,
mol wt   average Mw ~135,000 by LS
concentration   50 wt. % in ethanol
refractive index   n20/D 1.419
acid number   245‑275 mg KOH/g
density   0.977 g/mL at 25 °C

Description

Application

Functions in coatings, adhesives and cosmetics as a film former, emulsion stabilizer, controlled-release matrix, temporary protective coating and binding agent in powders.

Features and Benefits

Film former with good clarity, adhesion and moisture resistance. Acid group promotes adhesion to hair, paper, glass, metals and plastics.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1866 3/PG 2
WGK Germany 
3
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?