• USA Home
  • 416509 - Tetrapropenylsuccinic anhydride,mixture of isomers

EMAIL THIS PAGE TO A FRIEND
416509 Aldrich

Tetrapropenylsuccinic anhydride,mixture of isomers

≥90%

  • CAS Number 26544-38-7

  • Empirical Formula (Hill Notation) C16H26O3

  • Molecular Weight 266.38

  •  EC Number 247-781-6

Purchase

Properties

InChI Key   RYDRJNGBRZDNMR-MDWZMJQESA-N
assay   ≥90%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
WN1300000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?