• USA Home
  • 41729 - N,N-Dimethylundecylamine

EMAIL THIS PAGE TO A FRIEND
41729 Aldrich

N,N-Dimethylundecylamine

purum, ≥98.0% (GC)

Synonym: 1-(Dimethylamino)undecane

  • CAS Number 17373-28-3

  • Linear Formula CH3(CH2)10N(CH3)2

  • Molecular Weight 199.38

  •  Beilstein Registry Number 2345290

  •  MDL number MFCD00077748

  •  PubChem Substance ID 24866135

Purchase

Properties

Related Categories Amines, Building Blocks, C13, Chemical Synthesis, Nitrogen Compounds,
InChI Key   MMWFTWUMBYZIRZ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.435(lit.)
bp   50 °C/0.1 mmHg(lit.)
density   0.783 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
220 °F
Flash Point(C) 
104 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?